missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Cyanoethyl)glycine 98%, ACROS Organics™

Manufacturer:  Acros Organics 154900250

 View more versions of this product

Catalog No. AC154900250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-(2-Cyanoethyl)glycine
98%
97.5
Authentic
≥97.5%
C5H8N2O2
MFCD00066903
C(CNCC(=O)O)C#N
128.131
128.13
Powder or Flakes
25g
3088-42-4
100.0
Glass bottle
98%
NCCH2CH2NHCH2CO2H
KZUBZCHAWPDYQX-UHFFFAOYSA-N
2-(2-cyanoethylamino)acetic acid
76526
White
188°C to 193°C

Chemical Identifiers

3088-42-4
128.131
KZUBZCHAWPDYQX-UHFFFAOYSA-N
C5H8N2O2
MFCD00066903
76526
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-418-1

SDS Documents