Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Chloroethyl)acetamide, 97%

$38.61 - $157.05

Chemical Identifiers

Chemical Name or Material N- (2-Chloroethyl)acetamide
CAS 7355-58-0
Molecular Formula CH3CONHCH2CH2Cl
Formula Weight 121.57g/mol
MDL Number MFCD00000965
Beilstein 1743108
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1180209

 
alfa aesar™
A11802-09
10g Each for $38.61

AAA1180218

 
alfa aesar™
A11802-18
50g Each for $157.05
Description & Specifications

Specifications

Quantity 10g
Boiling Point 130° to 132°C (11mmHg)
Quantity 10g
Flash Point >110°C (230°F)
Chemical Name or Material N- (2-Chloroethyl)acetamide
CAS 7355-58-0
Assay 97%
Molecular Formula CH3CONHCH2CH2Cl
Formula Weight 121.57g/mol
MDL Number MFCD00000965
Beilstein 1743108
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev
RTECS Number AB5460000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.