Please login to your online account to display your discounted pricing

N-(2-Chloroacetyl)-L-tyrosine, 99%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  225780010

 View more versions of this product

Catalog No. AC225780010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Powder
Formula Weight 257.66
Quantity 1g
Color White
Beilstein 14, 614
EINECS Number 214-544-3
CAS Max % 100.0
Molecular Formula C11H12ClNO4
Infrared Spectrum Authentic
Name Note 99%
Linear Formula 4-(HO)C6H4CH2CH(NHCOCH2Cl)CO2H
CAS 1145-56-8
Chemical Name or Material N-(2-Chloroacetyl)-L-tyrosine
Assay Percent Range 99%
CAS Min % 98.5
MDL Number MFCD00002390
Specific Rotation +57° to +61° (20°C, 589nm) (c=2, C2H5OH)

Specifications

Chemical Name or Material N-(2-Chloroacetyl)-L-tyrosine
Name Note 99%
CAS 1145-56-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H12ClNO4
Linear Formula 4-(HO)C6H4CH2CH(NHCOCH2Cl)CO2H
Formula Weight 257.66
MDL Number MFCD00002390