Please login to your online account to display your discounted pricing

N-(2-Chloroacetyl)-L-tyrosine, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material N-(2-Chloroacetyl)-L-tyrosine
Name Note 99%
CAS 1145-56-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H12ClNO4
Linear Formula 4-(HO)C6H4CH2CH(NHCOCH2Cl)CO2H
Formula Weight 257.66
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC225780050

  SDS
thermo fisher scientific
225780050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC225780010

  SDS
thermo fisher scientific
225780010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Powder
Formula Weight 257.66
Quantity 5g
Color White
EINECS Number 214-544-3
Beilstein 14, 614
CAS Max % 100.0
Molecular Formula C11H12ClNO4
CAS 1145-56-8
CAS Min % 98.5
Name Note 99%
Chemical Name or Material N-(2-Chloroacetyl)-L-tyrosine
Linear Formula 4-(HO)C6H4CH2CH(NHCOCH2Cl)CO2H
MDL Number MFCD00002390
Assay Percent Range 99%
Specific Rotation +57° to +61° (20°C, 589nm) (c=2, C2H5OH)
Infrared Spectrum Authentic