Please login to your online account to display your discounted pricing

Alfa Aesar™ N-[2-Chloro-5-(trifluoromethyl)phenyl]thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-[2-Chloro-5-(trifluoromethyl)phenyl]thiourea
CAS 21714-35-2
Molecular Formula C8H6ClF3N2S
Formula Weight 254.66
MDL Number MFCD00041155
UN Number UN2811
Beilstein 2735893
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2224803

 
alfa aesar™
B2224803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2224806

 
alfa aesar™
B2224806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.66
UN Number UN2811
Health Hazard 1 Danger
Chemical Name or Material N-[2-Chloro-5-(trifluoromethyl)phenyl]thiourea
Assay 97%
Molecular Formula C8H6ClF3N2S
Melting Point 140° to 142°C
MDL Number MFCD00041155
CAS 21714-35-2
Beilstein 2735893
Quantity 1g
Odor Odorless
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.