Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Chloro-3-pyridyl)maleimide

$194.38 - $867.89

Chemical Identifiers

Chemical Name or Material N-(2-Chloro-3-pyridyl)maleimide
CAS 278610-39-2
Molecular Formula C9H5ClN2O2
Formula Weight 208.61
MDL Number MFCD00111928
Synonym N-(2-Chloropyridin-3-yl)maleimide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5054703

  SDS
alfa aesar™
H5054703
1g Each for $194.38

AAH5054706

  SDS
alfa aesar™
H5054706
5g Each for $867.89
Description & Specifications

Specifications

Formula Weight 208.61
Health Hazard 1 Warning
CAS 278610-39-2
Molecular Formula C9H5ClN2O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00111928
Synonym N-(2-Chloropyridin-3-yl)maleimide
Chemical Name or Material N-(2-Chloro-3-pyridyl)maleimide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.