Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Carboxyphenyl)phthalimide, 98%

$211.26 - $211.26

Chemical Identifiers

Chemical Name or Material N-(2-Carboxyphenyl)phthalimide
CAS 41513-78-4
Molecular Formula C15H9NO4
Formula Weight 267.24g/mol
MDL Number MFCD00041218
Beilstein 232897
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1027514

 
alfa aesar™
L10275-14
25g Each for $211.26
Description & Specifications

Specifications

Quantity 25g
Melting Point 218° to 220°C
Quantity 25g
Chemical Name or Material N-(2-Carboxyphenyl)phthalimide
CAS 41513-78-4
Assay 98%
Molecular Formula C15H9NO4
Formula Weight 267.24g/mol
MDL Number MFCD00041218
Beilstein 232897
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.