missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Carboxyphenyl)glycine, 97%, ACROS Organics™

$103.83 - $103.83

Chemical Identifiers

CAS 612-42-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H9NO4
Linear Formula HOCC6H4NHCHCOH
Formula Weight 195.17
MDL Number MFCD00020445
Chemical Name or Material N-(2-Carboxyphenyl)glycine
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC305130010 SDS Thermo Fisher Scientific
AC305130010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC305130050 SDS Thermo Fisher Scientific
305130050
Glass bottle 5g Each for $103.83
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

612-42-0
100.0
C9H9NO4
195.17
N-(2-Carboxyphenyl)glycine
96.0
97%
HOCC6H4NHCHCOH
MFCD00020445
≥96.0%

Specifications

195.17
1g
100.0
Glass bottle
97%
MFCD00020445
HOCC6H4NHCHCOH
≥96.0%
Crystalline Powder
Light yellow to yellow
96.0
612-42-0
C9H9NO4
Authentic
N-(2-Carboxyphenyl)glycine