missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Carboxyphenyl)glycine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 305130050

 View more versions of this product

Catalog No. AC305130050


 

Specifications Description & Specifications

Specifications

N-(2-Carboxyphenyl)glycine
5g
96.0
Authentic
≥96.0%
Glass bottle
HOCC6H4NHCHCOH
MFCD00020445
Light yellow to yellow
612-42-0
100.0
Crystalline Powder
97%
C9H9NO4
195.17

Chemical Identifiers

612-42-0
100.0
C9H9NO4
195.17
N-(2-Carboxyphenyl)glycine
96.0
97%
HOCC6H4NHCHCOH
MFCD00020445
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.