Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Carboxyphenyl)glycine, 97%

$32.78 - $82.35

Chemical Identifiers

Chemical Name or Material N-(2-Carboxyphenyl)glycine
CAS 612-42-0
Molecular Formula C9H9NO4
Formula Weight 195.17
MDL Number MFCD00020445
Synonym 2-(Carboxymethylamino)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2770103

 
alfa aesar™
H2770103
1g Each for $32.78

AAH2770106

 
alfa aesar™
H2770106
5g Each for $82.35
Description & Specifications

Specifications

Formula Weight 195.17
Chemical Name or Material N-(2-Carboxyphenyl)glycine
TSCA TSCA
Assay 97%
Molecular Formula C9H9NO4
MDL Number MFCD00020445
CAS 612-42-0
Synonym 2-(Carboxymethylamino)benzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point ≈218°C (dec.)
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.