missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Carboxyethyl)iminodiacetic Acid, TCI America™

$94.72 - $259.73

Chemical Identifiers

CAS 6245-75-6
Molecular Formula C7H11NO6
Molecular Weight (g/mol) 205.166
MDL Number MFCD00059634
InChI Key UWRBFYBQPCJRRL-UHFFFAOYSA-N
Synonym n-2-carboxyethyl iminodiacetic acid, nitrilodiacetic-propionic acid, beta-alanine diacetic acid, unii-kou355f9z2, n,n-bis carboxymethyl-, a-alanine, .beta.-alanine, n,n-bis carboxymethyl, 3-bis carboxymethyl amino propanoic acid, n-2-carboxyethyl dimethylsulfonium chloride, n,n-bis carboxymethyl-beta-alanine, beta-alaninediacetic acid
PubChem CID 535795
IUPAC Name 3-[bis(carboxymethyl)amino]propanoic acid
SMILES C(CN(CC(=O)O)CC(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N040825G View Documents TCI America
N040825G
25g Each for $259.73
 
N04085G View Documents TCI America
N04085G
5g Each for $94.72
 
Specifications

Chemical Identifiers

6245-75-6
205.166
UWRBFYBQPCJRRL-UHFFFAOYSA-N
535795
C(CN(CC(=O)O)CC(=O)O)C(=O)O
C7H11NO6
MFCD00059634
n-2-carboxyethyl iminodiacetic acid, nitrilodiacetic-propionic acid, beta-alanine diacetic acid, unii-kou355f9z2, n,n-bis carboxymethyl-, a-alanine, .beta.-alanine, n,n-bis carboxymethyl, 3-bis carboxymethyl amino propanoic acid, n-2-carboxyethyl dimethylsulfonium chloride, n,n-bis carboxymethyl-beta-alanine, beta-alaninediacetic acid
3-[bis(carboxymethyl)amino]propanoic acid

Specifications

White
25g
C7H11NO6
n-2-carboxyethyl iminodiacetic acid, nitrilodiacetic-propionic acid, beta-alanine diacetic acid, unii-kou355f9z2, n,n-bis carboxymethyl-, a-alanine, .beta.-alanine, n,n-bis carboxymethyl, 3-bis carboxymethyl amino propanoic acid, n-2-carboxyethyl dimethylsulfonium chloride, n,n-bis carboxymethyl-beta-alanine, beta-alaninediacetic acid
C(CN(CC(=O)O)CC(=O)O)C(=O)O
205.166
205.17
≥98.0% (T)
195°C
6245-75-6
MFCD00059634
UWRBFYBQPCJRRL-UHFFFAOYSA-N
3-[bis(carboxymethyl)amino]propanoic acid
535795
Crystal-Powder at 20°C
N-(2-Carboxyethyl)iminodiacetic Acid
Safety and Handling

Safety and Handling

TSCA : Yes