missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Butynyl)phthalimide, 97%

Manufacturer:  Alfa Aesar™ H5350914

 View more versions of this product

Catalog No. AAH5350914



Specifications

Specifications

N-(2-Butynyl)phthalimide
25g
C12H9NO2
1-phthalimido-2-butyne
CC#CCN1C(=O)C2=CC=CC=C2C1=O
199.209
199.21
219°C to 223°C
113439-83-1
MFCD00160830
COPWPECHFWIYSP-UHFFFAOYSA-N
2-but-2-ynylisoindole-1,3-dione
8063352
97%

Chemical Identifiers

113439-83-1
199.209
COPWPECHFWIYSP-UHFFFAOYSA-N
8063352
CC#CCN1C(=O)C2=CC=CC=C2C1=O
C12H9NO2
MFCD00160830
1-phthalimido-2-butyne
2-but-2-ynylisoindole-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents