missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Bromophenyl)-4-nitrobenzamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 99514-85-9
Molecular Formula C13H9BrN2O3
Molecular Weight (g/mol) 321.13
MDL Number MFCD00475065
InChI Key FRFOXBSTPQRRJL-UHFFFAOYSA-N
Synonym n-2-bromophenyl-4-nitrobenzamide, cambridge id 5273912, 4-nitrobenzamide, n-2-bromophenyl, n-2-bromo-phenyl-4-nitro-benzamide, n-2-bromophenyl 4-nitrophenyl carboxamide
PubChem CID 2056203
IUPAC Name N-(2-bromophenyl)-4-nitrobenzamide
SMILES C1=CC=C(C(=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60711MD View Documents Alfa Aesar™
H60711MD
250mg Each for $220.53
 
AAH6071103 View Documents Alfa Aesar™
H6071103
1g Each for $703.20
 
Specifications

Chemical Identifiers

99514-85-9
321.13
FRFOXBSTPQRRJL-UHFFFAOYSA-N
2056203
C1=CC=C(C(=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])Br
C13H9BrN2O3
MFCD00475065
n-2-bromophenyl-4-nitrobenzamide, cambridge id 5273912, 4-nitrobenzamide, n-2-bromophenyl, n-2-bromo-phenyl-4-nitro-benzamide, n-2-bromophenyl 4-nitrophenyl carboxamide
N-(2-bromophenyl)-4-nitrobenzamide

Specifications

250mg
C13H9BrN2O3
2749484
FRFOXBSTPQRRJL-UHFFFAOYSA-N
N-(2-bromophenyl)-4-nitrobenzamide
2056203
97%
99514-85-9
MFCD00475065
n-2-bromophenyl-4-nitrobenzamide, cambridge id 5273912, 4-nitrobenzamide, n-2-bromophenyl, n-2-bromo-phenyl-4-nitro-benzamide, n-2-bromophenyl 4-nitrophenyl carboxamide
C1=CC=C(C(=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])Br
321.13
321.13
N-(2-Bromophenyl)-4-nitrobenzamide