Please login to your online account to display your discounted pricing

N-(2-Bromoethyl)phthalimide ≥99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC106975000

 View more versions of this product

Catalog No. AC106975000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 254.08
Assay ≥96.0% (on dry substance)
Physical Form Crystalline Powder
Quantity 500g
Color White to slightly pink or beige
TSCA TSCA
EINECS Number 209-379-9
Melting Point 81° to 84°C
CAS Max % 100.0
CAS Min % 96.0
Water 1% max. (K.F.)
Infrared Spectrum Authentic
Molecular Formula C10H8BrNO2
Assay Percent Range 97%
Chemical Name or Material N-(2-Bromoethyl)phthalimide
CAS 574-98-1
MDL Number MFCD00005902
Name Note 99+%
Packaging Glass bottle

Specifications

Chemical Name or Material N-(2-Bromoethyl)phthalimide
Name Note 99+%
CAS 574-98-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (on dry substance)
Assay Percent Range 97%
Molecular Formula C10H8BrNO2
Formula Weight 254.08
MDL Number MFCD00005902