missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals
$76.47 - $842.08
Chemical Identifiers
CAS | 574-98-1 |
---|---|
Molecular Formula | C10H8BrNO2 |
Molecular Weight (g/mol) | 254.083 |
MDL Number | MFCD00005902 |
InChI Key | CHZXTOCAICMPQR-UHFFFAOYSA-N |
Synonym | n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dione |
PubChem CID | 11325 |
IUPAC Name | 2-(2-bromoethyl)isoindole-1,3-dione |
SMILES | C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1216414
|
Thermo Scientific Chemicals
A1216414 |
25 g |
Each for $76.47
|
|
|||||
AAA1216422
|
Thermo Scientific Chemicals
A1216422 |
100 g |
Each for $204.53
|
|
|||||
AAA1216436
|
Thermo Scientific Chemicals
A1216436 |
500 g |
Each for $842.08
|
|
|||||
Description
N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.
Solubility
Soluble in water (224 mg/L at 25°C).
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Chemical Identifiers
574-98-1 | |
254.083 | |
CHZXTOCAICMPQR-UHFFFAOYSA-N | |
11325 | |
C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr |
C10H8BrNO2 | |
MFCD00005902 | |
n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dione | |
2-(2-bromoethyl)isoindole-1,3-dione |
Specifications
574-98-1 | |
C10H8BrNO2 | |
25 g | |
n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dione | |
CHZXTOCAICMPQR-UHFFFAOYSA-N | |
2-(2-bromoethyl)isoindole-1,3-dione | |
11325 | |
≥98% |
80°C to 84°C | |
MFCD00005902 | |
148736 | |
Soluble in water (224mg/L at 25°C). | |
C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr | |
254.083 | |
254.09 | |
N- (2-Bromoethyl)phthalimide |
Safety and Handling
EINECSNumber : 209-379-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only