Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Bromobenzyloxycarbonyloxy)succinimide, 98+%

Chemical Identifiers

Chemical Name or Material N-(2-Bromobenzyloxycarbonyloxy)succinimide
CAS 128611-93-8
Molecular Formula C12H10BrNO5
Formula Weight 328.13
MDL Number MFCD00082848
UN Number UN3335
Beilstein 3623239
Synonym 2-Bromobenzyl succinimidyl carbonate; Z(2-Br)-OSu
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1347106

 
alfa aesar™
L1347106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1347114

 
alfa aesar™
L1347114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 328.13
Chemical Name or Material N-(2-Bromobenzyloxycarbonyloxy)succinimide
Assay ≥98%
CAS 128611-93-8
Molecular Formula C12H10BrNO5
Melting Point 105° to 108°C
MDL Number MFCD00082848
Quantity 5g
Synonym 2-Bromobenzyl succinimidyl carbonate; Z(2-Br)-OSu
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
UN Number UN3335
Beilstein 3623239
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.

2° to 8°C