missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Bromobenzyloxycarbonyloxy)succinimide, 98+%

Manufacturer:  Alfa Aesar™ L1347114

 View more versions of this product

Catalog No. AAL1347114



Specifications

Specifications

N-(2-Bromobenzyloxycarbonyloxy)succinimide
25g
C12H10BrNO5
UN3335
n-2-bromobenzyloxycarbonyloxy succinimide, z 2-br-osu, carbonic acid 2-bromobenzyl succinimidyl ester, 2-bromobenzyl succinimidyl carbonate, 2-bromobenzyl succinimdyl carbonate, n-2-bromobenzyloxycarbonyloxy succinimide, 2,5-dioxoazolidinyl 2-bromophenyl methoxy formate, z 2-br-osu, n-2-bromobenzyloxycarbonyloxy succinimide, 2-bromophenyl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 2,5-pyrrolidinedione, 1-2-bromophenyl methoxy carbonyl oxy
C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2Br
328.118
328.13
105°C to 108°C
128611-93-8
MFCD00082848
3623239
SZDNRTVADOCWKU-UHFFFAOYSA-N
(2-bromophenyl)methyl (2,5-dioxopyrrolidin-1-yl) carbonate
598169
≥98%

Chemical Identifiers

128611-93-8
328.118
SZDNRTVADOCWKU-UHFFFAOYSA-N
598169
C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2Br
C12H10BrNO5
MFCD00082848
n-2-bromobenzyloxycarbonyloxy succinimide, z 2-br-osu, carbonic acid 2-bromobenzyl succinimidyl ester, 2-bromobenzyl succinimidyl carbonate, 2-bromobenzyl succinimdyl carbonate, n-2-bromobenzyloxycarbonyloxy succinimide, 2,5-dioxoazolidinyl 2-bromophenyl methoxy formate, z 2-br-osu, n-2-bromobenzyloxycarbonyloxy succinimide, 2-bromophenyl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 2,5-pyrrolidinedione, 1-2-bromophenyl methoxy carbonyl oxy
(2-bromophenyl)methyl (2,5-dioxopyrrolidin-1-yl) carbonate
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.

Recommended Storage : 2° to 8°C

Documents