Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Bromobenzyl)acetamide, 97%

$197.03 - $858.92

Chemical Identifiers

Chemical Name or Material N-(2-Bromobenzyl)acetamide
CAS 74315-07-4
Molecular Formula C9H10BrNO
Formula Weight 228.09
MDL Number MFCD00831169
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5059903

 
alfa aesar™
H5059903
1g Each for $197.03

AAH5059906

 
alfa aesar™
H5059906
5g Each for $858.92
Description & Specifications

Specifications

Formula Weight 228.09
Chemical Name or Material N-(2-Bromobenzyl)acetamide
Assay 97%
Molecular Formula C9H10BrNO
Melting Point 78° to 80°C
MDL Number MFCD00831169
CAS 74315-07-4
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.