Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Bromo-4-methylphenyl)thiourea, 98%

Chemical Identifiers

Chemical Name or Material N-(2-Bromo-4-methylphenyl)thiourea
CAS 66644-79-9
Molecular Formula C8H9BrN2S
Formula Weight 245.15
MDL Number MFCD00041147
UN Number UN2811
Beilstein 2719238
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1177403

  SDS
alfa aesar™
L1177403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1177406

  SDS
alfa aesar™
L1177406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.15
Chemical Name or Material N-(2-Bromo-4-methylphenyl)thiourea
CAS 66644-79-9
Assay 98%
Molecular Formula C8H9BrN2S
Melting Point 173° to 176°C
MDL Number MFCD00041147
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN2811
Beilstein 2719238
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.