missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Bromo-4-methylphenyl)thiourea, 98%

Manufacturer:  Alfa Aesar™ L1177406

 View more versions of this product

Catalog No. AAL1177406



Specifications

Specifications

N-(2-Bromo-4-methylphenyl)thiourea
5g
C8H9BrN2S
UN2811
1-2-bromo-4-methylphenyl-2-thiourea, 2-bromo-4-methylphenyl thiourea, n-2-bromo-4-methylphenyl thiourea, 1-2-bromo-4-methylphenyl thiourea, 2-bromo-4-methylphenyl thiourea, 2-bromo-4-methylphenyl-thiourea, 2-bromo-4-methyl-phenyl-thiourea
CC1=CC(=C(C=C1)NC(=S)N)Br
245.138
245.15
173°C to 176°C
66644-79-9
MFCD00041147
2719238
QBTARKBNALVKLA-UHFFFAOYSA-N
(2-bromo-4-methylphenyl)thiourea
4011538
98%

Chemical Identifiers

66644-79-9
245.138
QBTARKBNALVKLA-UHFFFAOYSA-N
4011538
CC1=CC(=C(C=C1)NC(=S)N)Br
C8H9BrN2S
MFCD00041147
1-2-bromo-4-methylphenyl-2-thiourea, 2-bromo-4-methylphenyl thiourea, n-2-bromo-4-methylphenyl thiourea, 1-2-bromo-4-methylphenyl thiourea, 2-bromo-4-methylphenyl thiourea, 2-bromo-4-methylphenyl-thiourea, 2-bromo-4-methyl-phenyl-thiourea
(2-bromo-4-methylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents