missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-[2-Benzyl-2-(Boc-amino)ethyl]isoindole-1,3-dione, 97%

Manufacturer:  Alfa Aesar™ H6398103

 View more versions of this product

Catalog No. AAH6398103



Specifications

Specifications

N-[2-Benzyl-2-(Boc-amino)ethyl]isoindole-1,3-dione
885266-56-8
MFCD03844710
ZBCKKSQEEBLGDU-UHFFFAOYSA-N
tert-butyl N-[1-(1,3-dioxoisoindol-2-yl)-3-phenylpropan-2-yl]carbamate
24720887
Solid
1g
C22H24N2O4
tert-butyl 1-benzyl-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethylcarbamate, tert-butyl n-1-1,3-dioxoisoindol-2-yl-3-phenylpropan-2-yl carbamate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CN2C(=O)C3=CC=CC=C3C2=O
380.444
380.44
97%

Chemical Identifiers

885266-56-8
380.444
ZBCKKSQEEBLGDU-UHFFFAOYSA-N
24720887
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CN2C(=O)C3=CC=CC=C3C2=O
C22H24N2O4
MFCD03844710
tert-butyl 1-benzyl-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethylcarbamate, tert-butyl n-1-1,3-dioxoisoindol-2-yl-3-phenylpropan-2-yl carbamate
tert-butyl N-[1-(1,3-dioxoisoindol-2-yl)-3-phenylpropan-2-yl]carbamate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents