missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[2-Benzyl-2-(Boc-amino)ethyl]isoindole-1,3-dione, 97%, Alfa Aesar™

$383.99 - $1,225.99

Chemical Identifiers

CAS 885266-56-8
Molecular Formula C22H24N2O4
Molecular Weight (g/mol) 380.444
MDL Number MFCD03844710
InChI Key ZBCKKSQEEBLGDU-UHFFFAOYSA-N
Synonym tert-butyl 1-benzyl-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethylcarbamate, tert-butyl n-1-1,3-dioxoisoindol-2-yl-3-phenylpropan-2-yl carbamate
PubChem CID 24720887
IUPAC Name tert-butyl N-[1-(1,3-dioxoisoindol-2-yl)-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CN2C(=O)C3=CC=CC=C3C2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63981MD View Documents Alfa Aesar
H63981MD
250mg Each for $383.99
 
AAH6398103 View Documents Alfa Aesar
H6398103
1g Each for $1,225.99
 
Specifications

Chemical Identifiers

885266-56-8
380.444
ZBCKKSQEEBLGDU-UHFFFAOYSA-N
24720887
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CN2C(=O)C3=CC=CC=C3C2=O
C22H24N2O4
MFCD03844710
tert-butyl 1-benzyl-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethylcarbamate, tert-butyl n-1-1,3-dioxoisoindol-2-yl-3-phenylpropan-2-yl carbamate
tert-butyl N-[1-(1,3-dioxoisoindol-2-yl)-3-phenylpropan-2-yl]carbamate

Specifications

250mg
C22H24N2O4
tert-butyl 1-benzyl-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethylcarbamate, tert-butyl n-1-1,3-dioxoisoindol-2-yl-3-phenylpropan-2-yl carbamate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CN2C(=O)C3=CC=CC=C3C2=O
380.444
380.44
97%
885266-56-8
MFCD03844710
ZBCKKSQEEBLGDU-UHFFFAOYSA-N
tert-butyl N-[1-(1,3-dioxoisoindol-2-yl)-3-phenylpropan-2-yl]carbamate
24720887
Solid
N-[2-Benzyl-2-(Boc-amino)ethyl]isoindole-1,3-dione
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C