missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Aminoethyl)pyrrolidine, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC103701000

 View more versions of this product

Catalog No. AC103701000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-(2-Aminoethyl)pyrrolidine
Authentic
1.4677 to 1.4697 (20°C, 589nm)
100mL
98.5
99%
MFCD00003182
WRXNJTBODVGDRY-UHFFFAOYSA-N
2-pyrrolidin-1-ylethanamine
1344
Liquid
0.9g/mL
Glass bottle
Colorless to Brown
7154-73-6
100.0
C6H14N2
1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine
C1CCN(C1)CCN
114.192
114.19
≥98.5%

Chemical Identifiers

7154-73-6
114.192
WRXNJTBODVGDRY-UHFFFAOYSA-N
1344
C1CCN(C1)CCN
C6H14N2
MFCD00003182
1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine
2-pyrrolidin-1-ylethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents