Please login to your online account to display your discounted pricing

N-(2-Aminoethyl)acetamide, 90%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC223170250

 View more versions of this product

Catalog No. AC223170250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 102.14
Quantity 25g
Beilstein 04, III, 522
CAS Min % 88.0
CAS Max % 100.0
Chemical Name or Material N-(2-Aminoethyl)acetamide
Molecular Formula C4H10N2O
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 1001-53-2
Assay Percent Range 90%
MDL Number MFCD00008163
Packaging Glass bottle
Linear Formula CH3C(O)NH(CH2)2NH2
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material N-(2-Aminoethyl)acetamide
CAS 1001-53-2
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C4H10N2O
Linear Formula CH3C(O)NH(CH2)2NH2
Formula Weight 102.14
MDL Number MFCD00008163
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents