Please login to your online account to display your discounted pricing

Alfa Aesar™ N(2),9-Diacetylguanine, 99%

$73.97 - $73.97

Chemical Identifiers

Chemical Name or Material N(2),9-Diacetylguanine
CAS 3056-33-5
Molecular Formula C9H9N5O3
Formula Weight 235.2
MDL Number MFCD00142116
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2270906

  SDS
alfa aesar™
B2270906
5g Each for $73.97
Description & Specifications

Specifications

Formula Weight 235.2
Health Hazard 1 Warning
Quantity 5g
Assay 99%
Melting Point 284° to 286°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00142116
CAS 3056-33-5
Chemical Name or Material N(2),9-Diacetylguanine
Recommended Storage −30° to −10°C
Molecular Formula C9H9N5O3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C