missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dimethylphenyl)thiourea, 99%

Manufacturer:  Alfa Aesar™ L1299803

 View more versions of this product

Catalog No. AAL1299803



Specifications

Specifications

N-(2,6-Dimethylphenyl)thiourea
~195°C
6396-76-5
MFCD00041165
2209836
ASNKJUONFPQYPC-UHFFFAOYSA-N
(2,6-dimethylphenyl)thiourea
853911
99%
Odorless
1g
C9H12N2S
UN2811
1-2,6-dimethylphenyl thiourea, 1-2,6-dimethylphenyl-2-thiourea, 2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl thiourea, 1-2,6-xylyl thiourea, thiourea, 2,6-dimethylphenyl, amino 2,6-dimethylphenyl amino methane-1-thione, acmc-20apd5, 1-2,6-xylyl-2-thiourea, 2,6-dimethyl-phenyl-thiourea
CC1=C(C(=CC=C1)C)NC(=S)N
180.269
180.27

Chemical Identifiers

6396-76-5
180.269
ASNKJUONFPQYPC-UHFFFAOYSA-N
853911
CC1=C(C(=CC=C1)C)NC(=S)N
C9H12N2S
MFCD00041165
1-2,6-dimethylphenyl thiourea, 1-2,6-dimethylphenyl-2-thiourea, 2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl thiourea, 1-2,6-xylyl thiourea, thiourea, 2,6-dimethylphenyl, amino 2,6-dimethylphenyl amino methane-1-thione, acmc-20apd5, 1-2,6-xylyl-2-thiourea, 2,6-dimethyl-phenyl-thiourea
(2,6-dimethylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents