missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dimethylphenyl)-4-methylbenzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H55784MD

 View more versions of this product

Catalog No. AAH55784MD



Specifications

Specifications

N-(2,6-Dimethylphenyl)-4-methylbenzenesulfonamide
4703-15-5
MFCD00185176
SEYRJIDJWCOIBS-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)-4-methylbenzenesulfonamide
262999
97%
250mg
C15H17NO2S
1-p-toluenesulfonamido-2,6-dimethylbenzene
CC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC=C2C)C
275.366
275.37

Chemical Identifiers

4703-15-5
275.366
SEYRJIDJWCOIBS-UHFFFAOYSA-N
262999
CC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC=C2C)C
C15H17NO2S
MFCD00185176
1-p-toluenesulfonamido-2,6-dimethylbenzene
N-(2,6-dimethylphenyl)-4-methylbenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents