missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dimethylphenyl)-4-methylbenzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H5578403

 View more versions of this product

Catalog No. AAH5578403



Specifications

Specifications

N-(2,6-Dimethylphenyl)-4-methylbenzenesulfonamide
4703-15-5
MFCD00185176
SEYRJIDJWCOIBS-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)-4-methylbenzenesulfonamide
262999
97%
1g
C15H17NO2S
1-p-toluenesulfonamido-2,6-dimethylbenzene
CC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC=C2C)C
275.366
275.37

Chemical Identifiers

4703-15-5
275.366
SEYRJIDJWCOIBS-UHFFFAOYSA-N
262999
CC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC=C2C)C
C15H17NO2S
MFCD00185176
1-p-toluenesulfonamido-2,6-dimethylbenzene
N-(2,6-dimethylphenyl)-4-methylbenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents