missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dimethylphenyl)-1-piperazineacetamide, 95%

Manufacturer:  Alfa Aesar™ H6185903

 View more versions of this product

Catalog No. AAH6185903



Specifications

Specifications

N-(2,6-Dimethylphenyl)-1-piperazineacetamide
1g
C14H21N3O
n-2,6-dimethylphenyl-2-piperazin-1-yl acetamide, n-2,6-diphenylmethyl-1-piperazine acetylamine, n-2,6-dimethylphenyl-1-piperazineacetamide, n-2,6-dimethylphenyl-2-piperazin-1-ylacetamide, unii-nys3i6283h, 1-piperazineacetamide, n-2,6-dimethylphenyl, 1-n-2,6-dimethylphenyl carbamoylmethyl piperazine, n-2,6-dimethylphenyl-2-piperazinylacetamide, n-2,6-dimethylphenyl-2-piperazin-1-yl-acetamide, 4-2,6-dimethylphenyl aminocarbonylmethyl piperazine
CC1=C(C(=CC=C1)C)NC(=O)CN2CCNCC2
247.342
247.34
95%
Gray
5294-61-1
MFCD06384973
NJKRFQIWDJSYOK-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide
2608606
Crystalline Powder

Chemical Identifiers

5294-61-1
247.342
NJKRFQIWDJSYOK-UHFFFAOYSA-N
2608606
CC1=C(C(=CC=C1)C)NC(=O)CN2CCNCC2
C14H21N3O
MFCD06384973
n-2,6-dimethylphenyl-2-piperazin-1-yl acetamide, n-2,6-diphenylmethyl-1-piperazine acetylamine, n-2,6-dimethylphenyl-1-piperazineacetamide, n-2,6-dimethylphenyl-2-piperazin-1-ylacetamide, unii-nys3i6283h, 1-piperazineacetamide, n-2,6-dimethylphenyl, 1-n-2,6-dimethylphenyl carbamoylmethyl piperazine, n-2,6-dimethylphenyl-2-piperazinylacetamide, n-2,6-dimethylphenyl-2-piperazin-1-yl-acetamide, 4-2,6-dimethylphenyl aminocarbonylmethyl piperazine
N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents