missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dimethylphenyl)-1-piperazineacetamide, 95%

$45.47 - $533.59

Chemical Identifiers

CAS 5294-61-1
Molecular Formula C14H21N3O
Molecular Weight (g/mol) 247.342
MDL Number MFCD06384973
InChI Key NJKRFQIWDJSYOK-UHFFFAOYSA-N
Synonym 1-n-2,6-dimethylphenyl carbamoylmethyl piperazine
PubChem CID 2608606
IUPAC Name N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)CN2CCNCC2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61859MD View Documents Alfa Aesar™
H61859MD
250mg Each for $45.47
 
AAH6185903 View Documents Alfa Aesar™
H6185903
1g Each for $134.53
 
AAH6185906 View Documents Alfa Aesar™
H6185906
5g Each for $533.59
 
Specifications

Chemical Identifiers

5294-61-1
247.342
NJKRFQIWDJSYOK-UHFFFAOYSA-N
2608606
CC1=C(C(=CC=C1)C)NC(=O)CN2CCNCC2
C14H21N3O
MFCD06384973
1-n-2,6-dimethylphenyl carbamoylmethyl piperazine
N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide

Specifications

Gray
5294-61-1
MFCD06384973
NJKRFQIWDJSYOK-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide
2608606
Crystalline Powder
N-(2,6-Dimethylphenyl)-1-piperazineacetamide
250mg
C14H21N3O
1-n-2,6-dimethylphenyl carbamoylmethyl piperazine
CC1=C(C(=CC=C1)C)NC(=O)CN2CCNCC2
247.342
247.34
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures