missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dimethylphenyl)-1-piperazineacetamide, 95%

Manufacturer:  Alfa Aesar™ H61859MD

 View more versions of this product

Catalog No. AAH61859MD



Specifications

Specifications

N-(2,6-Dimethylphenyl)-1-piperazineacetamide
250mg
C14H21N3O
1-n-2,6-dimethylphenyl carbamoylmethyl piperazine, 1-piperazineacetamide, n-2,6-dimethylphenyl, 4-2,6-dimethylphenyl aminocarbonylmethyl piperazine, n-2,6-dimethylphenyl-1-piperazineacetamide, n-2,6-dimethylphenyl-2-piperazin-1-yl acetamide, n-2,6-dimethylphenyl-2-piperazin-1-yl-acetamide, n-2,6-dimethylphenyl-2-piperazin-1-ylacetamide, n-2,6-dimethylphenyl-2-piperazinylacetamide, n-2,6-diphenylmethyl-1-piperazine acetylamine, unii-nys3i6283h
CC1=C(C(=CC=C1)C)NC(=O)CN2CCNCC2
247.342
247.34
95%
Gray
5294-61-1
MFCD06384973
NJKRFQIWDJSYOK-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide
2608606
Crystalline Powder

Chemical Identifiers

5294-61-1
247.342
NJKRFQIWDJSYOK-UHFFFAOYSA-N
2608606
CC1=C(C(=CC=C1)C)NC(=O)CN2CCNCC2
C14H21N3O
MFCD06384973
1-n-2,6-dimethylphenyl carbamoylmethyl piperazine, 1-piperazineacetamide, n-2,6-dimethylphenyl, 4-2,6-dimethylphenyl aminocarbonylmethyl piperazine, n-2,6-dimethylphenyl-1-piperazineacetamide, n-2,6-dimethylphenyl-2-piperazin-1-yl acetamide, n-2,6-dimethylphenyl-2-piperazin-1-yl-acetamide, n-2,6-dimethylphenyl-2-piperazin-1-ylacetamide, n-2,6-dimethylphenyl-2-piperazinylacetamide, n-2,6-diphenylmethyl-1-piperazine acetylamine, unii-nys3i6283h
N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents