missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Difluorophenyl)-4-methoxybenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 155997-19-6
Molecular Formula C14H11F2NO2
Molecular Weight (g/mol) 263.244
MDL Number MFCD04310051
InChI Key NDUMEFRGYXBRDG-UHFFFAOYSA-N
Synonym n-2,6-difluorophenyl-4-methoxybenzamide, chembl45855, n-2,6-difluoro-phenyl-4-methoxy-benzamide, n-2,6-difluorophenyl 4-methoxyphenyl carboxamide
PubChem CID 956606
IUPAC Name N-(2,6-difluorophenyl)-4-methoxybenzamide
SMILES COC1=CC=C(C=C1)C(=O)NC2=C(C=CC=C2F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59959MD View Documents Alfa Aesar™
H59959MD
250mg Each for $211.27
 
AAH5995903 View Documents Alfa Aesar™
H5995903
1g Each for $675.45
 
Specifications

Chemical Identifiers

155997-19-6
263.244
NDUMEFRGYXBRDG-UHFFFAOYSA-N
956606
COC1=CC=C(C=C1)C(=O)NC2=C(C=CC=C2F)F
C14H11F2NO2
MFCD04310051
n-2,6-difluorophenyl-4-methoxybenzamide, chembl45855, n-2,6-difluoro-phenyl-4-methoxy-benzamide, n-2,6-difluorophenyl 4-methoxyphenyl carboxamide
N-(2,6-difluorophenyl)-4-methoxybenzamide

Specifications

250mg
C14H11F2NO2
n-2,6-difluorophenyl-4-methoxybenzamide, chembl45855, n-2,6-difluoro-phenyl-4-methoxy-benzamide, n-2,6-difluorophenyl 4-methoxyphenyl carboxamide
COC1=CC=C(C=C1)C(=O)NC2=C(C=CC=C2F)F
263.244
263.24
N-(2,6-Difluorophenyl)-4-methoxybenzamide
155997-19-6
MFCD04310051
NDUMEFRGYXBRDG-UHFFFAOYSA-N
N-(2,6-difluorophenyl)-4-methoxybenzamide
956606
97%