Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dichlorophenyl)thiourea, 99%

Chemical Identifiers

Chemical Name or Material N-(2,6-Dichlorophenyl)thiourea
CAS 6590-91-6
Molecular Formula C7H6Cl2N2S
Formula Weight 221.11
MDL Number MFCD00041156
UN Number UN2811
Beilstein 2212144
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1811706

  SDS
alfa aesar™
A1811706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1811714

  SDS
alfa aesar™
A1811714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.11
Chemical Name or Material N-(2,6-Dichlorophenyl)thiourea
Assay 99%
Molecular Formula C7H6Cl2N2S
Melting Point 155° to 159°C
MDL Number MFCD00041156
CAS 6590-91-6
UN Number UN2811
Beilstein 2212144
Quantity 5g
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.