missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dichlorophenyl)benzenesulfonamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 77167-06-7
Molecular Formula C12H9Cl2NO2S
Molecular Weight (g/mol) 302.169
MDL Number MFCD02007023
InChI Key CQELSZQJFRLUFQ-UHFFFAOYSA-N
Synonym n-2,6-dichlorophenyl benzenesulfonamide
PubChem CID 12723885
IUPAC Name N-(2,6-dichlorophenyl)benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC=C2Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60316MD View Documents Alfa Aesar™
H60316MD
250mg Each for $220.53
 
AAH6031603 View Documents Alfa Aesar™
H6031603
1g Each for $703.20
 
Specifications

Chemical Identifiers

77167-06-7
302.169
CQELSZQJFRLUFQ-UHFFFAOYSA-N
12723885
C1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC=C2Cl)Cl
C12H9Cl2NO2S
MFCD02007023
n-2,6-dichlorophenyl benzenesulfonamide
N-(2,6-dichlorophenyl)benzenesulfonamide

Specifications

250mg
C12H9Cl2NO2S
3099420
CQELSZQJFRLUFQ-UHFFFAOYSA-N
N-(2,6-dichlorophenyl)benzenesulfonamide
12723885
97%
77167-06-7
MFCD02007023
n-2,6-dichlorophenyl benzenesulfonamide
C1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC=C2Cl)Cl
302.169
302.17
N-(2,6-Dichlorophenyl)benzenesulfonamide