missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,6-Dichlorophenyl)-2-methoxybenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 362602-54-8
Molecular Formula C14H11Cl2NO2
Molecular Weight (g/mol) 296.147
MDL Number MFCD01115478
InChI Key FVOARQOPPNATSM-UHFFFAOYSA-N
Synonym n-2,6-dichlorophenyl-2-methoxybenzamide, n-2,6-dichlorophenyl 2-methoxyphenyl carboxamide
PubChem CID 4392873
IUPAC Name N-(2,6-dichlorophenyl)-2-methoxybenzamide
SMILES COC1=CC=CC=C1C(=O)NC2=C(C=CC=C2Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59796MD View Documents Alfa Aesar™
H59796MD
250mg Each for $211.27
 
AAH5979603 View Documents Alfa Aesar™
H5979603
1g Each for $675.45
 
Specifications

Chemical Identifiers

362602-54-8
296.147
FVOARQOPPNATSM-UHFFFAOYSA-N
4392873
COC1=CC=CC=C1C(=O)NC2=C(C=CC=C2Cl)Cl
C14H11Cl2NO2
MFCD01115478
n-2,6-dichlorophenyl-2-methoxybenzamide, n-2,6-dichlorophenyl 2-methoxyphenyl carboxamide
N-(2,6-dichlorophenyl)-2-methoxybenzamide

Specifications

250mg
C14H11Cl2NO2
n-2,6-dichlorophenyl-2-methoxybenzamide, n-2,6-dichlorophenyl 2-methoxyphenyl carboxamide
COC1=CC=CC=C1C(=O)NC2=C(C=CC=C2Cl)Cl
296.147
296.15
N-(2,6-Dichlorophenyl)-2-methoxybenzamide
362602-54-8
MFCD01115478
FVOARQOPPNATSM-UHFFFAOYSA-N
N-(2,6-dichlorophenyl)-2-methoxybenzamide
4392873
97%