Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,5-Dimethoxyphenyl)thiourea, 99%

Chemical Identifiers

Chemical Name or Material N-(2,5-Dimethoxyphenyl)thiourea
CAS 67617-98-5
Molecular Formula C9H12N2O2S
Formula Weight 212.27
MDL Number MFCD00022163
UN Number UN2811
Beilstein 2727068
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1145403

  SDS
alfa aesar™
L1145403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1145406

  SDS
alfa aesar™
L1145406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.27
Chemical Name or Material N-(2,5-Dimethoxyphenyl)thiourea
Molecular Formula C9H12N2O2S
MDL Number MFCD00022163
CAS 67617-98-5
UN Number UN2811
Health Hazard 1 Danger
Assay 99%
Melting Point 162° to 165°C
Beilstein 2727068
Quantity 1g
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.