missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,5-Dimethoxyphenyl)thiourea, 99%

Manufacturer:  Alfa Aesar™ L1145403

 View more versions of this product

Catalog No. AAL1145403



Specifications

Specifications

N-(2,5-Dimethoxyphenyl)thiourea
1g
C9H12N2O2S
UN2811
2,5-dimethoxyphenyl thiourea, 1-2,5-dimethoxyphenyl-2-thiourea, 1-2,5-dimethoxyphenyl thiourea, amino 2,5-dimethoxyphenyl amino methane-1-thione, 2,5-dimethoxy phenyl thiourea, n-2,5-dimethoxyphenyl thiourea, 2,5-dimethoxy-phenyl-thiourea
COC1=CC(=C(C=C1)OC)NC(=S)N
212.267
212.27
162°C to 165°C
67617-98-5
MFCD00022163
2727068
ZLGQKHQZDDTIFW-UHFFFAOYSA-N
(2,5-dimethoxyphenyl)thiourea
2758483
99%

Chemical Identifiers

67617-98-5
212.267
ZLGQKHQZDDTIFW-UHFFFAOYSA-N
2758483
COC1=CC(=C(C=C1)OC)NC(=S)N
C9H12N2O2S
MFCD00022163
2,5-dimethoxyphenyl thiourea, 1-2,5-dimethoxyphenyl-2-thiourea, 1-2,5-dimethoxyphenyl thiourea, amino 2,5-dimethoxyphenyl amino methane-1-thione, 2,5-dimethoxy phenyl thiourea, n-2,5-dimethoxyphenyl thiourea, 2,5-dimethoxy-phenyl-thiourea
(2,5-dimethoxyphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents