Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,5-Difluorophenyl)thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-(2,5-Difluorophenyl)thiourea
CAS 207981-44-0
Molecular Formula C7H6F2N2S
Formula Weight 188.2
MDL Number MFCD00041152
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1222303

  SDS
alfa aesar™
L1222303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1222306

  SDS
alfa aesar™
L1222306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.2
Molecular Formula C7H6F2N2S
Chemical Name or Material N-(2,5-Difluorophenyl)thiourea
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
CAS 207981-44-0
Health Hazard 1 Danger
Assay 97%
UN Number UN2811
Quantity 1g
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
MDL Number MFCD00041152
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 138° to 142°C
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.