missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,5-Difluorophenyl)thiourea, 97%

Manufacturer:  Alfa Aesar™ L1222303

 View more versions of this product

Catalog No. AAL1222303



Specifications

Specifications

N-(2,5-Difluorophenyl)thiourea
1g
C7H6F2N2S
UN2811
JBWSNSWRTOFDBM-UHFFFAOYSA-N
(2,5-difluorophenyl)thiourea
2774223
97%
138°C to 142°C
207981-44-0
MFCD00041152
1-2,5-difluorophenyl thiourea, 1-2,5-difluorophenyl-2-thiourea, 2,5-difluorophenyl thiourea, n-2,5-difluorophenyl thiourea, amino 2,5-difluorophenyl amino methane-1-thione, acmc-209w9g, 2,5-difluoro-phenyl-thiourea, n-2,5-difluorophenyl-thiourea, thiourea,n-2,5-difluorophenyl, 2,5-difluorophenyl thiourea
C1=CC(=C(C=C1F)NC(=S)N)F
188.196
188.2

Chemical Identifiers

207981-44-0
188.196
JBWSNSWRTOFDBM-UHFFFAOYSA-N
2774223
C1=CC(=C(C=C1F)NC(=S)N)F
C7H6F2N2S
MFCD00041152
1-2,5-difluorophenyl thiourea, 1-2,5-difluorophenyl-2-thiourea, 2,5-difluorophenyl thiourea, n-2,5-difluorophenyl thiourea, amino 2,5-difluorophenyl amino methane-1-thione, acmc-209w9g, 2,5-difluoro-phenyl-thiourea, n-2,5-difluorophenyl-thiourea, thiourea,n-2,5-difluorophenyl, 2,5-difluorophenyl thiourea
(2,5-difluorophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents