Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,5-Dichlorophenyl)thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-(2,5-Dichlorophenyl)thiourea
CAS 4949-85-3
Molecular Formula C7H6Cl2N2S
Formula Weight 221.11
MDL Number MFCD00041154
UN Number UN2811
Beilstein 2212146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1198403

 
alfa aesar™
L1198403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1198406

 
alfa aesar™
L1198406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.11
Chemical Name or Material N-(2,5-Dichlorophenyl)thiourea
CAS 4949-85-3
Assay 97%
Molecular Formula C7H6Cl2N2S
MDL Number MFCD00041154
Beilstein 2212146
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 192° to 195°C
UN Number UN2811
Quantity 1g
Odor Odorless
RTECS Number YS9275000
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.