missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,5-Dichlorophenyl)thiourea, 97%

Manufacturer:  Alfa Aesar™ L1198403

 View more versions of this product

Catalog No. AAL1198403



Specifications

Specifications

N-(2,5-Dichlorophenyl)thiourea
192°C to 195°C
4949-85-3
MFCD00041154
2212146
UQCOANNCPUOZNW-UHFFFAOYSA-N
(2,5-dichlorophenyl)thiourea
2731095
97%
Odorless
1g
C7H6Cl2N2S
UN2811
1-2,5-dichlorophenyl thiourea, n-2,5-dichlorophenyl thiourea, 2,5-dichlorophenyl thiourea, thiourea, 2,5-dichlorophenyl, 2,5-dichlorophenyl thiourea, 1-2,5-dichlorophenyl-2-thiourea, 2,5-dichloro-phenyl-thiourea, urea, 1-2,5-dichlorophenyl-2-thio, thiourea, 2,5-dichlorophenyl-9ci, amino 2,5-dichlorophenyl amino methane-1-thione
C1=CC(=C(C=C1Cl)NC(=S)N)Cl
221.099
221.11

Chemical Identifiers

4949-85-3
221.099
UQCOANNCPUOZNW-UHFFFAOYSA-N
2731095
C1=CC(=C(C=C1Cl)NC(=S)N)Cl
C7H6Cl2N2S
MFCD00041154
1-2,5-dichlorophenyl thiourea, n-2,5-dichlorophenyl thiourea, 2,5-dichlorophenyl thiourea, thiourea, 2,5-dichlorophenyl, 2,5-dichlorophenyl thiourea, 1-2,5-dichlorophenyl-2-thiourea, 2,5-dichloro-phenyl-thiourea, urea, 1-2,5-dichlorophenyl-2-thio, thiourea, 2,5-dichlorophenyl-9ci, amino 2,5-dichlorophenyl amino methane-1-thione
(2,5-dichlorophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : YS9275000

Documents