Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,5-Dichlorophenyl)maleamic acid, 97%

$113.62 - $455.04

Chemical Identifiers

Chemical Name or Material N-(2,5-Dichlorophenyl)maleamic acid
CAS 31460-32-9
Molecular Formula C10H7Cl2NO3
Formula Weight 260.07
MDL Number MFCD00082906
Synonym 4-(2,5-Dichloroanilino)-4-oxo-2-butenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2534206

  SDS
alfa aesar™
B2534206
5g Each for $113.62

AAB2534214

  SDS
alfa aesar™
B2534214
25g Each for $455.04
Description & Specifications

Specifications

Formula Weight 260.07
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Melting Point 140° to 144°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material N-(2,5-Dichlorophenyl)maleamic acid
Synonym 4-(2,5-Dichloroanilino)-4-oxo-2-butenoic acid
Assay 97%
Molecular Formula C10H7Cl2NO3
CAS 31460-32-9
MDL Number MFCD00082906
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.