missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,5-Dichlorophenyl)-4-methoxybenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 7465-94-3
Molecular Formula C14H11Cl2NO2
Molecular Weight (g/mol) 296.147
MDL Number MFCD00541840
InChI Key XQSFQIDFRWNBTM-UHFFFAOYSA-N
Synonym n-2,5-dichlorophenyl-4-methoxybenzamide, n-2,5-dichloro-phenyl-4-methoxy-benzamide, benzamide, n-2,5-dichlorophenyl-4-methoxy, n-2,5-dichlorophenyl-4-methoxybenzamide
PubChem CID 346041
IUPAC Name N-(2,5-dichlorophenyl)-4-methoxybenzamide
SMILES COC1=CC=C(C=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59876MD View Documents Alfa Aesar™
H59876MD
250mg Each for $211.27
 
AAH5987603 View Documents Alfa Aesar™
H5987603
1g Each for $675.45
 
Specifications

Chemical Identifiers

7465-94-3
296.147
XQSFQIDFRWNBTM-UHFFFAOYSA-N
346041
COC1=CC=C(C=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl
C14H11Cl2NO2
MFCD00541840
n-2,5-dichlorophenyl-4-methoxybenzamide, n-2,5-dichloro-phenyl-4-methoxy-benzamide, benzamide, n-2,5-dichlorophenyl-4-methoxy, n-2,5-dichlorophenyl-4-methoxybenzamide
N-(2,5-dichlorophenyl)-4-methoxybenzamide

Specifications

250mg
C14H11Cl2NO2
n-2,5-dichlorophenyl-4-methoxybenzamide, n-2,5-dichloro-phenyl-4-methoxy-benzamide, benzamide, n-2,5-dichlorophenyl-4-methoxy, n-2,5-dichlorophenyl-4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl
296.147
296.15
N-(2,5-Dichlorophenyl)-4-methoxybenzamide
7465-94-3
MFCD00541840
XQSFQIDFRWNBTM-UHFFFAOYSA-N
N-(2,5-dichlorophenyl)-4-methoxybenzamide
346041
97%