Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4-Dimethylphenyl)thiourea, 98%

Chemical Identifiers

Chemical Name or Material N-(2,4-Dimethylphenyl)thiourea
CAS 16738-20-8
Molecular Formula C9H12N2S
Formula Weight 180.27
MDL Number MFCD00041164
UN Number UN2811
Beilstein 2639862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1220603

  SDS
alfa aesar™
L1220603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1220606

  SDS
alfa aesar™
L1220606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.27
Chemical Name or Material N-(2,4-Dimethylphenyl)thiourea
CAS 16738-20-8
Assay 98%
Health Hazard 3 GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Beilstein 2639862
MDL Number MFCD00041164
Molecular Formula C9H12N2S
Melting Point 177° to 180°C
Odor Odorless
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
RTECS Number YU3675000
Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300
Fatal if swallowed.