missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4-Dimethylphenyl)thiourea, 98%

Manufacturer:  Alfa Aesar™ L1220606

 View more versions of this product

Catalog No. AAL1220606



Specifications

Specifications

N-(2,4-Dimethylphenyl)thiourea
177°C to 180°C
16738-20-8
MFCD00041164
2639862
KAGLPYRXTCQWHU-UHFFFAOYSA-N
(2,4-dimethylphenyl)thiourea
2798617
98%
Odorless
5g
C9H12N2S
UN2811
1-2,4-dimethylphenyl-2-thiourea, 2,4-dimethylphenyl thiourea, 2,4-xylylthiourea, n-2,4-dimethylphenyl thiourea, 1-2,4-dimethylphenyl thiourea, 2,4-xylyl thiourea, 2-thio-1-2,4-xylyl urea, urea, 2-thio-1-2,4-xylyl, thiourea, 2,4-dimethylphenyl-9ci, thiourea,n-2,4-dimethylphenyl
CC1=CC(=C(C=C1)NC(=S)N)C
180.269
180.27

Chemical Identifiers

16738-20-8
180.269
KAGLPYRXTCQWHU-UHFFFAOYSA-N
2798617
CC1=CC(=C(C=C1)NC(=S)N)C
C9H12N2S
MFCD00041164
1-2,4-dimethylphenyl-2-thiourea, 2,4-dimethylphenyl thiourea, 2,4-xylylthiourea, n-2,4-dimethylphenyl thiourea, 1-2,4-dimethylphenyl thiourea, 2,4-xylyl thiourea, 2-thio-1-2,4-xylyl urea, urea, 2-thio-1-2,4-xylyl, thiourea, 2,4-dimethylphenyl-9ci, thiourea,n-2,4-dimethylphenyl
(2,4-dimethylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : YU3675000

Documents