Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4-Dimethoxyphenyl)thiourea, 99%

Chemical Identifiers

Chemical Name or Material N-(2,4-Dimethoxyphenyl)thiourea
CAS 35696-77-6
Molecular Formula C9H12N2O2S
Formula Weight 212.27
MDL Number MFCD00041158
UN Number UN2811
Beilstein 2109660
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0964603

  SDS
alfa aesar™
L0964603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0964606

  SDS
alfa aesar™
L0964606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.27
Chemical Name or Material N-(2,4-Dimethoxyphenyl)thiourea
MDL Number MFCD00041158
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay 99%
Molecular Formula C9H12N2O2S
CAS 35696-77-6
UN Number UN2811
Quantity 1g
Health Hazard 1 Danger
Melting Point 190° to 192°C
Beilstein 2109660
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.