Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4-Dimethoxyphenyl)thiourea, 99%


Quantity 5g

Manufacturer: alfa aesar™  L0964606

 View more versions of this product

Catalog No. AAL0964606

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 212.27
Chemical Name or Material N-(2,4-Dimethoxyphenyl)thiourea
Assay 99%
Molecular Formula C9H12N2O2S
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
CAS 35696-77-6
MDL Number MFCD00041158
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 1 Danger
Melting Point 190° to 192°C
Quantity 5g
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Beilstein 2109660
UN Number UN2811

Specifications

Chemical Name or Material N-(2,4-Dimethoxyphenyl)thiourea
CAS 35696-77-6
Molecular Formula C9H12N2O2S
Formula Weight 212.27
MDL Number MFCD00041158
UN Number UN2811
Beilstein 2109660
Accessories

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.

SDS & Safety Documents