missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4-Dimethoxyphenyl)-4-nitrobenzamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 152586-90-8
Molecular Formula C15H14N2O5
Molecular Weight (g/mol) 302.286
MDL Number MFCD00406462
InChI Key XUJKRQHILJWOBB-UHFFFAOYSA-N
Synonym n-2,4-dimethoxyphenyl-4-nitrobenzamide, cambridge id 5107184, 4-nitro-n-2,4-dimethoxyphenyl benzamide, n-2,4-dimethoxy-phenyl-4-nitro-benzamide, n-2,4-dimethoxyphenyl 4-nitrophenyl carboxamide
PubChem CID 710798
IUPAC Name N-(2,4-dimethoxyphenyl)-4-nitrobenzamide
SMILES COC1=CC(=C(C=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60379MD View Documents Alfa Aesar™
H60379MD
250mg Each for $220.53
 
AAH6037903 View Documents Alfa Aesar™
H6037903
1g Each for $703.20
 
Specifications

Chemical Identifiers

152586-90-8
302.286
XUJKRQHILJWOBB-UHFFFAOYSA-N
710798
COC1=CC(=C(C=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])OC
C15H14N2O5
MFCD00406462
n-2,4-dimethoxyphenyl-4-nitrobenzamide, cambridge id 5107184, 4-nitro-n-2,4-dimethoxyphenyl benzamide, n-2,4-dimethoxy-phenyl-4-nitro-benzamide, n-2,4-dimethoxyphenyl 4-nitrophenyl carboxamide
N-(2,4-dimethoxyphenyl)-4-nitrobenzamide

Specifications

250mg
C15H14N2O5
7544958
XUJKRQHILJWOBB-UHFFFAOYSA-N
N-(2,4-dimethoxyphenyl)-4-nitrobenzamide
710798
97%
152586-90-8
MFCD00406462
n-2,4-dimethoxyphenyl-4-nitrobenzamide, cambridge id 5107184, 4-nitro-n-2,4-dimethoxyphenyl benzamide, n-2,4-dimethoxy-phenyl-4-nitro-benzamide, n-2,4-dimethoxyphenyl 4-nitrophenyl carboxamide
COC1=CC(=C(C=C1)NC(=O)C2=CC=C(C=C2)[N+](=O)[O-])OC
302.286
302.28
N-(2,4-Dimethoxyphenyl)-4-nitrobenzamide