missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2,4-Difluorophenyl)thiourea, 97%, Thermo Scientific™

$40.23 - $133.00

Chemical Identifiers

CAS 175277-76-6
Molecular Formula C7H6F2N2S
Molecular Weight (g/mol) 188.196
MDL Number MFCD00041150
InChI Key DZZSKQFBAGZNSH-UHFFFAOYSA-N
Synonym 1-2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl-2-thiourea, 2,4-difluoro-phenyl-thiourea, 2,4-difluorophenyl thiourea, acmc-1bxvl, amino 2,4-difluorophenyl amino methane-1-thione, buttpark 12118-31, difluorophenylthioacetonitrile, n-2,4-difluorophenyl thiourea, thiourea,n-2,4-difluorophenyl
PubChem CID 2734207
IUPAC Name (2,4-difluorophenyl)thiourea
SMILES C1=CC(=C(C=C1F)F)NC(=S)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1694503
View Documents Promotion Details
Thermo Fisher Scientific
A1694503
1g
Each for $40.23
 
AAA1694506
View Documents Promotion Details
Thermo Fisher Scientific
A1694506
5g
Each for $133.00
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

175277-76-6
188.196
DZZSKQFBAGZNSH-UHFFFAOYSA-N
2734207
C1=CC(=C(C=C1F)F)NC(=S)N
C7H6F2N2S
MFCD00041150
1-2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl-2-thiourea, 2,4-difluoro-phenyl-thiourea, 2,4-difluorophenyl thiourea, acmc-1bxvl, amino 2,4-difluorophenyl amino methane-1-thione, buttpark 12118-31, difluorophenylthioacetonitrile, n-2,4-difluorophenyl thiourea, thiourea,n-2,4-difluorophenyl
(2,4-difluorophenyl)thiourea

Specifications

175277-76-6
C7H6F2N2S
UN2811
1-2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl-2-thiourea, 2,4-difluoro-phenyl-thiourea, 2,4-difluorophenyl thiourea, acmc-1bxvl, amino 2,4-difluorophenyl amino methane-1-thione, buttpark 12118-31, difluorophenylthioacetonitrile, n-2,4-difluorophenyl thiourea, thiourea,n-2,4-difluorophenyl
C1=CC(=C(C=C1F)F)NC(=S)N
188.196
188.2
N-(2,4-Difluorophenyl)thiourea
165°C to 168°C
MFCD00041150
1g
DZZSKQFBAGZNSH-UHFFFAOYSA-N
(2,4-difluorophenyl)thiourea
2734207
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications