Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4-Dichlorophenyl)thiourea, 98+%

Chemical Identifiers

Chemical Name or Material N-(2,4-Dichlorophenyl)thiourea
CAS 6326-14-3
Molecular Formula C7H6Cl2N2S
Formula Weight 221.11
MDL Number MFCD00022161
UN Number UN2811
Beilstein 2096917
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1131406

 
alfa aesar™
L1131406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.11
Chemical Name or Material N-(2,4-Dichlorophenyl)thiourea
Assay ≥98%
Molecular Formula C7H6Cl2N2S
Melting Point 157° to 160°C
MDL Number MFCD00022161
CAS 6326-14-3
UN Number UN2811
Beilstein 2096917
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Odor Odorless
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Applications
N-(2,4-Dichlorophenyl)thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.